Gene Transcripts
- Gene Symbol
- Clns1a
- Entrez Gene ID
- 12729
- Gene Name
- chloride channel, nucleotide-sensitive, 1A
- Gene Aliases
- 2610036D06Rik, 2610100O04Rik, Clci, Clcni, ICLN
- Chromosome Location
- Chr. 7: 97696657 - 97720793 on Build GRCm38
- UniGene ID
- Mm.21482
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_023671.2 |
NP_076160.1
|
3, 4 |
454 |
|
GenBank mRNA |
AK011789.1 |
- |
3, 4 |
442 |
|
AK031877.1 |
- |
3, 4 |
436 |
|
AK078050.1 |
- |
3, 4 |
410 |
|
AK133981.1 |
- |
3, 4 |
447 |
|
AK151356.1 |
- |
3, 4 |
440 |
|
AK151944.1 |
- |
3, 4 |
440 |
|
AK169259.1 |
- |
3, 4 |
418 |
|
BC005575.1 |
- |
3, 4 |
420 |
3967933 |
CX236388.1 |
- |
3, 4 |
454 |
|
U53455.1 |
- |
3, 4 |
366 |
|
View Full Product Details