Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_010120.5 | NP_034250.3 | 3 | 574 | |
GenBank mRNA | AF026481.1 | - | 2 | 1393 | |
AK009026.1 | - | 3 | 545 | ||
AK013268.1 | - | 3 | 573 | ||
AK019451.1 | - | 3 | 575 | ||
AK150651.1 | - | 3 | 571 | ||
AK150673.1 | - | 3 | 547 | ||
AK150677.1 | - | 3 | 547 | ||
AK166368.1 | - | 546 | |||
AK167333.1 | - | 3 | 534 | ||
BC027437.1 | - | 3 | 536 | 4017054 | |
CT010335.1 | - | 1 | 221 |