Gene Transcripts

Gene Symbol
Ggps1
Entrez Gene ID
14593
Gene Name
geranylgeranyl diphosphate synthase 1
Gene Aliases
1810026C22Rik, 9530089B04Rik, AI843169, C79210
Chromosome Location
Chr. 13: 14049793 - 14063734 on Build GRCm38
UniGene ID
Mm.148039
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010282.2 NP_034412.1 4 719
GenBank mRNA AB016044.1 - 4 607
AK011980.1 - 5 864
AK132907.1 - 4 712
BC069913.1 - 4 719 6469191

View Full Product Details