Gene Transcripts

Gene Symbol
Lor
Entrez Gene ID
16939
Gene Name
loricrin
Gene Aliases
AI036317, S77319
Chromosome Location
Chr. 3: 92080271 - 92083142 on Build GRCm38
UniGene ID
Mm.1121
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008508.2 NP_032534.2 2 1729
GenBank mRNA BC026781.1 - 2 1742
BC058223.1 - 2 1717 6307099
M34398.1 - 2 1712

View Full Product Details