Gene Transcripts

Gene Symbol
Mpo
Entrez Gene ID
17523
Gene Name
myeloperoxidase
Gene Aliases
mKIAA4033
Chromosome Location
Chr. 11: 87793784 - 87804412 on Build GRCm38
UniGene ID
Mm.4668
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010824.2 NP_034954.2 4 631
GenBank mRNA AK160229.1 - 3 357
AK220229.1 - 3 404
AY500847.1 - 4 385
BC053912.1 - 3 351 30044601
CN661559.1 - 3 313
X15313.1 - 4 841

View Full Product Details