Gene Transcripts
- Gene Symbol
- ICAM3
- Entrez Gene ID
- 3385
- Gene Name
- intercellular adhesion molecule 3
- Gene Aliases
- CD50, CDW50, ICAM-R
- Chromosome Location
- Chr. 19: 10333776 - 10339834 on Build GRCh38
- UniGene ID
- Hs.654563
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001320605.1 |
NP_001307534.1
|
4, 5 |
1125 |
|
NM_001320606.1 |
NP_001307535.1
|
5, 6 |
1710 |
|
NM_001320608.1 |
NP_001307537.1
|
4, 5 |
1730 |
|
NM_002162.4 |
NP_002153.2
|
5, 6 |
1413 |
|
GenBank mRNA |
AF340060.1 |
- |
5, 6 |
1166 |
|
AF340061.1 |
- |
5, 6 |
1166 |
|
AK226023.1 |
- |
5, 6 |
1247 |
|
AK309643.1 |
- |
5, 6 |
1495 |
|
AK312826.1 |
- |
5, 6 |
1248 |
|
AX746321.1 |
- |
5, 6 |
1208 |
|
BC040123.1 |
- |
5, 6 |
1193 |
|
BC058903.1 |
- |
5, 6 |
1199 |
5207143 |
S50015.1 |
- |
5, 6 |
1194 |
|
X69711.1 |
- |
5, 6 |
1208 |
|
X69819.1 |
- |
5, 6 |
1201 |
|
View Full Product Details