Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001039132.2 | NP_001034221.1 | 3 | 758 | |
NM_001544.4 | NP_001535.1 | 3 | 835 | ||
NM_022377.3 | NP_071772.1 | 2 | 982 | ||
GenBank mRNA | BC000046.2 | - | 3 | 751 | |
BC029364.1 | - | 3 | 835 | 4659161 | |
BT009816.1 | - | 3 | 712 | ||
L27670.1 | - | 2 | 942 | ||
L27671.1 | - | 3 | 795 |