Gene Transcripts

Gene Symbol
Prkx
Entrez Gene ID
19108
Gene Name
protein kinase, X-linked
Gene Aliases
PKX1, Pkare
Chromosome Location
Chr. X: 77760972 - 77796288 on Build GRCm38
UniGene ID
Mm.290338
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016979.1 NP_058675.1 8 1267
GenBank mRNA AK034681.1 - 8 1586
AK037141.1 - 8 1261
AK081548.1 - 8 1247
AK139510.1 - 8 1275
AK149937.1 - 8 1245
AK150588.1 - 8 1229
AK152166.1 - 8 1063
AK154447.1 - 8 1247
AK169322.1 - 8 1218
BC006875.1 - 8 1267 3596013

View Full Product Details