Gene Transcripts

Gene Symbol
Ube2h
Entrez Gene ID
22214
Gene Name
ubiquitin-conjugating enzyme E2H
Gene Aliases
1500009C23Rik, AI181839, AI326965, AI462483, AW227540, E2-20K, Gid3, N28148, Ubc8
Chromosome Location
Chr. 6: 30211289 - 30304539 on Build GRCm38
UniGene ID
Mm.332967
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001169577.1 NP_001163048.1 3, 4 220
NM_009459.3 NP_033485.1 5, 6 737
GenBank mRNA AK131688.1 - 5, 6 715
BC008517.1 - 5, 6 543 3600233
U19854.1 - 5, 6 466

View Full Product Details