Gene Transcripts

Gene Symbol
Slx4
Entrez Gene ID
52864
Gene Name
SLX4 structure-specific endonuclease subunit homolog (S. cerevisiae)
Gene Aliases
AI256635, AI426760, Btbd12, D16Bwg1016e
Chromosome Location
Chr. 16: 3979105 - 4003829 on Build GRCm38
UniGene ID
Mm.29935
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177472.5 NP_803423.2 11 2739
GenBank mRNA AK173263.1 - 10 2152
BC065125.1 - 11 2744 30534878
BC082337.1 - 3 965

View Full Product Details