Gene Transcripts
- Gene Symbol
- Ndufa12
- Entrez Gene ID
- 66414
- Gene Name
- NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 12
- Gene Aliases
- 2410011G03Rik, AW112974
- Chromosome Location
- Chr. 10: 94199009 - 94220948 on Build GRCm38
- UniGene ID
- Mm.27886
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_025551.3 |
NP_079827.2
|
1, 2 |
130 |
|
GenBank mRNA |
AK003131.1 |
- |
1, 2 |
103 |
|
AK003322.1 |
- |
1, 2 |
103 |
|
AK008876.1 |
- |
1, 2 |
103 |
|
AK010460.1 |
- |
1, 2 |
131 |
|
AK032774.1 |
- |
1, 2 |
104 |
|
AK134765.1 |
- |
1, 2 |
103 |
|
AK153240.1 |
- |
1, 2 |
103 |
|
AK167943.1 |
- |
1, 2 |
96 |
|
BC004633.1 |
- |
|
1560 |
|
BC086922.1 |
- |
1, 2 |
135 |
6751147 |
View Full Product Details