Gene Transcripts

Gene Symbol
Ccdc77
Entrez Gene ID
67200
Gene Name
coiled-coil domain containing 77
Gene Aliases
2400002C23Rik, 2700091N06Rik, AV168274
Chromosome Location
Chr. 6: 120324322 - 120364369 on Build GRCm38
UniGene ID
Mm.45767
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026028.5 NP_080304.1 11 1422
GenBank mRNA AK012598.1 - 11 1314
AK031590.1 - 9 1226
BC018387.1 - 11 1250 3982552

View Full Product Details