Gene Transcripts

Gene Symbol
Dym
Entrez Gene ID
69190
Gene Name
dymeclin
Gene Aliases
1810041M12Rik, 4933427L07Rik, C030019K18Rik
Chromosome Location
Chr. 18: 75018699 - 75286966 on Build GRCm38
UniGene ID
Mm.173826
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027727.2 NP_082003.1 10 1378
GenBank mRNA AK049909.1 - 10 1343
AK140643.1 - 2 483
AK149628.1 - 10 1352
AK149879.1 - 10 1354
AK149891.1 - 10 1352
AK154794.1 - 10 1367
BC018220.1 - 6 738 3985017
BC038276.1 - 10 1380 4458792

View Full Product Details