Gene Transcripts

Gene Symbol
Lrrc63
Entrez Gene ID
70859
Gene Name
leucine rich repeat containing 63
Gene Aliases
4921509B22Rik
Chromosome Location
Chr. 14: 75084303 - 75130962 on Build GRCm38
UniGene ID
Mm.320289
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027581.1 NP_081857.1 10 2020
GenBank mRNA AK014850.1 - 9 1752
BC145796.1 - 10 2015 40131099

View Full Product Details