Gene Transcripts

Gene Symbol
Fmnl2
Entrez Gene ID
71409
Gene Name
formin-like 2
Gene Aliases
5430425K04Rik, Man
Chromosome Location
Chr. 2: 52857716 - 53134202 on Build GRCm38
UniGene ID
Mm.394427
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172409.2 NP_765997.2 6, 7 872
GenBank mRNA CA329111.1 - 2, 3 243
CJ044449.1 - 4, 5 375

View Full Product Details