Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001025594.1 | NP_001020765.1 | 3 | 1305 | |
NM_027947.2 | NP_082223.2 | 4 | 1397 | ||
GenBank mRNA | AK005818.1 | - | 4 | 1371 | |
AK122277.1 | - | 3 | 1276 | ||
AK147806.1 | - | 3 | 1300 | ||
BC043117.1 | - | 3 | 1305 | 6408745 |