Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_201226.1 | NP_957678.1 | 2 | 713 | |
GenBank mRNA | AK129306.1 | - | 2 | 522 | |
AK154897.1 | - | 2 | 743 | ||
BC094573.1 | - | 2 | 703 | 30354494 | |
BC138462.1 | - | 2 | 785 | 8861482 | |
BC138463.1 | - | 2 | 785 | 8861483 | |
BU148242.1 | - | 2 | 708 |