Gene Transcripts

Gene Symbol
Mcee
Entrez Gene ID
73724
Gene Name
methylmalonyl CoA epimerase
Gene Aliases
1110007A04Rik
Chromosome Location
Chr. 7: 64392612 - 64412121 on Build GRCm38
UniGene ID
Mm.10093
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028626.1 NP_082902.1 2 319
GenBank mRNA AK003511.1 - 2 316
BC038157.1 - 2 319 1479934

View Full Product Details