Gene Transcripts
- Gene Symbol
- Wdr33
- Entrez Gene ID
- 74320
- Gene Name
- WD repeat domain 33
- Gene Aliases
- 1110001N06Rik, 2310011G05Rik, 2810021O11Rik, 8430413N20Rik, WDC146
- Chromosome Location
- Chr. 18: 31804057 - 31908987 on Build GRCm38
- UniGene ID
- Mm.277705
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001170966.1 |
NP_001164437.1
|
2, 3 |
424 |
|
NM_001170967.1 |
NP_001164438.1
|
2, 3 |
424 |
|
NM_001170970.1 |
NP_001164441.1
|
2, 3 |
424 |
|
NM_028866.3 |
NP_083142.2
|
2, 3 |
424 |
|
GenBank mRNA |
AB086191.1 |
- |
1, 2 |
203 |
|
AK003256.1 |
- |
2, 3 |
392 |
|
AK009297.1 |
- |
2, 3 |
391 |
|
AK045125.1 |
- |
2, 3 |
384 |
|
AK045923.1 |
- |
2, 3 |
383 |
|
AK050653.1 |
- |
2, 3 |
384 |
|
AK078286.1 |
- |
2, 3 |
392 |
|
AK161982.1 |
- |
2, 3 |
384 |
|
BC011156.1 |
- |
2, 3 |
336 |
|
BC019463.1 |
- |
2, 3 |
424 |
|
BC030076.1 |
- |
2, 3 |
356 |
|
BC064745.1 |
- |
2, 3 |
305 |
|
View Full Product Details