Gene Transcripts

Gene Symbol
Krt76
Entrez Gene ID
77055
Gene Name
keratin 76
Gene Aliases
2310001L23Rik
Chromosome Location
Chr. 15: 101884351 - 101892920 on Build GRCm38
UniGene ID
Mm.195788
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033177.2 NP_001028349.1 6 1211
GenBank mRNA AK009075.1 - 6 1195
AK036802.1 - 6 1213
AK137676.1 - 6 1212

View Full Product Details