Gene Transcripts

Gene Symbol
Tm2d1
Entrez Gene ID
94043
Gene Name
TM2 domain containing 1
Gene Aliases
2310026L18Rik, AA990549, Bbp
Chromosome Location
Chr. 4: 98355370 - 98383281 on Build GRCm38
UniGene ID
Mm.331719
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053157.2 NP_444387.1 2 236
GenBank mRNA AF353993.1 - 2 210
AK009511.1 - 2 215
BC089492.1 - 2 236 6703812

View Full Product Details