Gene Transcripts
- Gene Symbol
- Eif3m
- Entrez Gene ID
- 98221
- Gene Name
- eukaryotic translation initiation factor 3, subunit M
- Gene Aliases
- Ga17, Pcid1, Tango7
- Chromosome Location
- Chr. 2: 104999656 - 105017027 on Build GRCm38
- UniGene ID
- Mm.379278
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_145380.2 |
NP_663355.1
|
7, 8 |
759 |
|
GenBank mRNA |
AK077480.1 |
- |
7, 8 |
760 |
|
AK084738.1 |
- |
7, 8 |
778 |
|
AK146222.1 |
- |
7, 8 |
791 |
|
AK168066.1 |
- |
7, 8 |
776 |
|
BC005598.1 |
- |
7, 8 |
759 |
3969335 |
BC050191.1 |
- |
1, 2 |
1491 |
|
BC103795.1 |
- |
7, 8 |
759 |
30135815 |
BC116787.1 |
- |
7, 8 |
756 |
40126106 |
BC116789.1 |
- |
7, 8 |
756 |
40126108 |
View Full Product Details