Gene Transcripts

Gene Symbol
Fbxl5
Entrez Gene ID
242960
Gene Name
F-box and leucine-rich repeat protein 5
Gene Aliases
Fbl4, Fir4
Chromosome Location
Chr. 5: 43744618 - 43782149 on Build GRCm38
UniGene ID
Mm.25794
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178729.4 NP_848844.1 9 2279
GenBank mRNA AK029957.1 - 9 2227
AK082931.1 - 9 2249
BC113798.1 - 9 2197 40062506

View Full Product Details