Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001568.2 | NP_001559.1 | 2 | 154 | |
XM_017013398.1 | XP_016868887.1 | 2 | 226 | ||
GenBank mRNA | AK124178.1 | - | 2 | 758 | |
AK310902.1 | - | 2 | 226 | ||
AK313186.1 | - | 2 | 143 | ||
BC000734.2 | - | 2 | 130 | 2959855 | |
BC005944.1 | - | 2 | 143 | ||
BC008419.1 | - | 2 | 148 | 4080571 | |
BC016706.1 | - | 2 | 145 | 4096434 | |
BC017887.1 | - | 2 | 150 | 4685684 | |
BC021679.1 | - | 2 | 145 | 4132737 | |
BG498300.1 | - | 2 | 155 | ||
CR457029.1 | - | 2 | 126 | ||
CR542275.1 | - | 2 | 126 | ||
KC333871.1 | - | 2 | 154 | ||
U54562.1 | - | 2 | 132 | ||
U62962.1 | - | 2 | 148 | ||
U85947.1 | - | 2 | 134 | ||
U94175.1 | - | 2 | 146 |