Gene Transcripts

Gene Symbol
Ndufa6
Entrez Gene ID
67130
Gene Name
NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 6 (B14)
Gene Aliases
14kDa, 2700038D15Rik, B230217P19Rik
Chromosome Location
Chr. 15: 82350139 - 82354291 on Build GRCm38
UniGene ID
Mm.27570
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025987.3 NP_080263.1 2 255
GenBank mRNA AK002749.1 - 2 202
AK010988.1 - 2 204
AK012328.1 - 2 204
BC048451.1 - 2 255 6773780

View Full Product Details