Gene Transcripts

Gene Symbol
Fzd7
Entrez Gene ID
14369
Gene Name
frizzled homolog 7 (Drosophila)
Gene Aliases
Fz7
Chromosome Location
Chr. 1: 59482147 - 59486955 on Build GRCm38
UniGene ID
Mm.297906
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008057.3 NP_032083.3 1 1112
GenBank mRNA BC049781.1 - 1 835
BC063077.1 - 2 443 6844727
U43320.1 - 1 660

View Full Product Details