Gene Transcripts

Gene Symbol
Vmn1r5
Entrez Gene ID
171192
Gene Name
vomeronasal 1 receptor 5
Gene Aliases
V1rc19
Chromosome Location
Chr. 6: 56985342 - 56986289 on Build GRCm38
UniGene ID
Mm.377172
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134174.2 NP_598935.2 1 271
GenBank mRNA BC138068.1 - 1 318
BC138069.1 - 1 318 8861088

View Full Product Details