Gene Transcripts

Gene Symbol
USP20
Entrez Gene ID
10868
Gene Name
ubiquitin specific peptidase 20
Gene Aliases
LSFR3A, VDU2, hVDU2
Chromosome Location
Chr. 9: 129835417 - 129881838 on Build GRCh38
UniGene ID
Hs.5452
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AB023220.1 - 4 303
AY074877.1 - 5 419
BC039593.1 - 5 329 5588842
DA791069.1 - 5 370

View Full Product Details