Gene Transcripts

Gene Symbol
CD69
Entrez Gene ID
969
Gene Name
CD69 molecule
Gene Aliases
AIM, BL-AC/P26, CLEC2C, EA1, GP32/28, MLR-3
Chromosome Location
Chr. 12: 9752486 - 9760901 on Build GRCh38
UniGene ID
Hs.208854
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001781.2 NP_001772.1 5 735
GenBank mRNA AK291869.1 - 5 735
BC007037.1 - 5 735 3950403
L07555.1 - 5 727
Z22576.1 - 5 735

View Full Product Details