Gene Transcripts

Gene Symbol
SPRY3
Entrez Gene ID
10251
Gene Name
sprouty RTK signaling antagonist 3
Gene Aliases
spry-3
Chromosome Location
Chr. X: 155612565 - 155782459 on Build GRCh38
UniGene ID
Hs.381912
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001304990.1 NP_001291919.1 3 8740
NM_005840.2 NP_005831.1 2 8884
GenBank mRNA BU164699.1 - 1 274

View Full Product Details