Gene Transcripts
- Gene Symbol
- EIF4A2
- Entrez Gene ID
- 1974
- Gene Name
- eukaryotic translation initiation factor 4A2
- Gene Aliases
- BM-010, DDX2B, EIF4A, EIF4F, eIF-4A-II, eIF4A-II
- Chromosome Location
- Chr. 3: 186783572 - 186789897 on Build GRCh38
- UniGene ID
- Hs.518475
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001967.3 |
NP_001958.2
|
3, 4 |
255 |
|
GenBank mRNA |
AK130087.1 |
- |
3, 4 |
250 |
|
AK225646.1 |
- |
3, 4 |
250 |
|
AY207392.1 |
- |
2, 3 |
186 |
|
BC012547.1 |
- |
3, 4 |
257 |
4079371 |
BC013708.1 |
- |
3, 4 |
235 |
4341420 |
BC015842.1 |
- |
3, 4 |
254 |
4337581 |
BC048105.1 |
- |
3, 4 |
253 |
4820774 |
BT009860.1 |
- |
3, 4 |
216 |
|
D30655.1 |
- |
3, 4 |
231 |
|
S79942.1 |
- |
3, 4 |
231 |
|
View Full Product Details