Gene Transcripts

Gene Symbol
STON1
Entrez Gene ID
11037
Gene Name
stonin 1
Gene Aliases
SALF, SBLF, STN1, STNB1
Chromosome Location
Chr. 2: 48530169 - 48598515 on Build GRCh38
UniGene ID
Hs.44385
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001198595.1 NP_001185524.1 5 4991
NM_006873.3 NP_006864.2 4 4874
GenBank mRNA AK023617.1 - 1 1743
CR936807.1 - 5 4974

View Full Product Details