Gene Transcripts
- Gene Symbol
- TRIM33
- Entrez Gene ID
- 51592
- Gene Name
- tripartite motif containing 33
- Gene Aliases
- ECTO, PTC7, RFG7, TF1G, TIF1G, TIF1GAMMA, TIFGAMMA
- Chromosome Location
- Chr. 1: 114392777 - 114511160 on Build GRCh38
- UniGene ID
- Hs.26837
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_015906.3 |
NP_056990.3
|
14, 15 |
2497 |
|
NM_033020.2 |
NP_148980.2
|
14, 15 |
2497 |
|
XM_005270936.3 |
XP_005270993.1
|
15, 16 |
2490 |
|
XM_005270937.3 |
XP_005270994.1
|
15, 16 |
2490 |
|
XM_011541568.2 |
XP_011539870.1
|
15, 16 |
2487 |
|
XM_017001452.1 |
XP_016856941.1
|
12, 13 |
1772 |
|
XM_017001453.1 |
XP_016856942.1
|
12, 13 |
1772 |
|
GenBank mRNA |
AB029036.1 |
- |
14, 15 |
2426 |
|
AF119043.1 |
- |
14, 15 |
2497 |
|
AF220136.1 |
- |
14, 15 |
2497 |
|
AF220137.1 |
- |
14, 15 |
2497 |
|
AJ132948.1 |
- |
14, 15 |
2188 |
|
AK021491.1 |
- |
1, 2 |
22 |
|
AK023434.1 |
- |
12, 13 |
1710 |
|
AK301997.1 |
- |
9, 10 |
1175 |
|
View Full Product Details