Gene Transcripts

Gene Symbol
GMPR2
Entrez Gene ID
51292
Gene Name
guanosine monophosphate reductase 2
Gene Aliases
GMPR 2
Chromosome Location
Chr. 14: 24232417 - 24239242 on Build GRCh38
UniGene ID
Hs.368855
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001002000.2 NP_001002000.1 10 1455
NM_001002001.2 NP_001002001.1 10 1396
NM_001002002.2 NP_001002002.1 10 1135
NM_001283021.1 NP_001269950.1 10 1714
NM_001283022.1 NP_001269951.1 8 1559
NM_001283023.1 NP_001269952.1 9 1051
NM_016576.4 NP_057660.2 9 1376
NR_104265.1 10 1479
XM_005267740.3 XP_005267797.1 9 1444
XM_005267741.3 XP_005267798.1 9 1503
XM_005267742.3 XP_005267799.1 9 1322
XM_006720165.2 XP_006720228.1 10 1422
XM_017021356.1 XP_016876845.1 10 1638
XM_017021357.1 XP_016876846.1 11 1494
XM_017021358.1 XP_016876847.1 11 1595
XM_017021359.1 XP_016876848.1 11 1238
XM_017021360.1 XP_016876849.1 7 1025
GenBank mRNA AB032903.1 - 9 1002
AF086919.1 - 10 1171
AF135159.1 - 10 1341
AF419346.1 - 10 1245
AK226166.1 - 10 1470
BC003053.1 - 8 1508 3546620
BC008021.1 - 9 1324 3537989
BC009832.2 - 10 1053 3958714
BC093039.1 - 10 1194 30347740
BX161436.1 - 10 1376
BX161518.1 - 10 1266
BX247993.1 - 9 974
CR457156.1 - 9 994

View Full Product Details