Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001290292.1 | NP_001277221.1 | 5 | 886 | |
NM_001290293.2 | NP_001277222.1 | 5 | 928 | ||
NM_001308075.1 | NP_001295004.1 | 5 | 848 | ||
NM_020195.2 | NP_064580.2 | 6 | 968 | ||
XM_011536975.1 | XP_011535277.1 | 6 | 1012 | ||
XM_011536976.1 | XP_011535278.1 | 3 | 1016 | ||
GenBank mRNA | AF226050.1 | - | 5 | 1242 | |
AK123513.1 | - | 4 | 2075 | ||
BC000989.2 | - | 6 | 968 | 2900140 | |
BC008566.1 | - | 5 | 1140 | ||
BX161384.1 | - | 4 | 879 |