Gene Transcripts

Gene Symbol
PYROXD2
Entrez Gene ID
84795
Gene Name
pyridine nucleotide-disulphide oxidoreductase domain 2
Gene Aliases
C10orf33, FP3420
Chromosome Location
Chr. 10: 98383565 - 98415221 on Build GRCh38
UniGene ID
Hs.238303
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK075265.1 - 3, 4 273
AK225637.1 - 3, 4 289
AK295699.1 - 3, 4 308
BC006131.1 - 3, 4 298 3626506
DC361966.1 - 3, 4 336

View Full Product Details