Gene Transcripts

Gene Symbol
SIRPD
Entrez Gene ID
128646
Gene Name
signal regulatory protein delta
Gene Aliases
PTPNS1L2, dJ576H24.4
Chromosome Location
Chr. 20: 1534251 - 1557697 on Build GRCh38
UniGene ID
Hs.729560
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178460.2 NP_848555.2 3, 4 629
GenBank mRNA AK093083.1 - 3, 4 1775
BC033502.1 - 3, 4 629 5172531

View Full Product Details