Gene Transcripts
- Gene Symbol
- PARP2
- Entrez Gene ID
- 10038
- Gene Name
- poly(ADP-ribose) polymerase 2
- Gene Aliases
- ADPRT2, ADPRTL2, ADPRTL3, ARTD2, PARP-2, pADPRT-2
- Chromosome Location
- Chr. 14: 20343041 - 20357904 on Build GRCh38
- UniGene ID
- Hs.409412
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001042618.1 |
NP_001036083.1
|
3, 4 |
309 |
|
NM_005484.3 |
NP_005475.2
|
3, 4 |
348 |
|
XM_005267247.3 |
XP_005267304.1
|
3, 4 |
348 |
|
XM_017020912.1 |
XP_016876401.1
|
3, 4 |
882 |
|
GenBank mRNA |
AF085734.1 |
- |
3, 4 |
345 |
|
AJ236876.2 |
- |
2, 3 |
259 |
|
AJ236912.1 |
- |
3, 4 |
283 |
|
AK001980.1 |
- |
3, 4 |
325 |
|
AK290079.1 |
- |
3, 4 |
288 |
|
AK300971.1 |
- |
3, 4 |
288 |
|
View Full Product Details