Gene Transcripts
- Gene Symbol
- OXCT2
- Entrez Gene ID
- 64064
- Gene Name
- 3-oxoacid CoA-transferase 2
- Gene Aliases
- FKSG25, SCOTT
- Chromosome Location
- Chr. 1: 39769525 - 39771348 on Build GRCh38
- UniGene ID
- Hs.472491
- Species
- Homo sapiens
- Gene Symbol
- OXCT2P1
- Entrez Gene ID
- 192217
- Gene Name
- 3-oxoacid CoA-transferase 2 pseudogene 1
- Gene Aliases
- OXCT2P
- Chromosome Location
- Chr. 1: 39514866 - 39516669 on Build GRCh38
- UniGene ID
- Hs.472491
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_132962.1 |
|
1 |
677 |
|
GenBank mRNA |
BC072388.1 |
- |
1 |
677 |
|
View Full Product Details