Gene Transcripts

Gene Symbol
LAG3
Entrez Gene ID
3902
Gene Name
lymphocyte activating 3
Gene Aliases
CD223
Chromosome Location
Chr. 12: 6772483 - 6778455 on Build GRCh38
UniGene ID
Hs.409523
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002286.5 NP_002277.4 6 1529
GenBank mRNA A21353.1 - 6 1410
AK292104.1 - 6 1503
X51985.3 - 6 1410

View Full Product Details