Gene Transcripts

Gene Symbol
ITIH2
Entrez Gene ID
3698
Gene Name
inter-alpha-trypsin inhibitor heavy chain 2
Gene Aliases
H2P, SHAP
Chromosome Location
Chr. 10: 7703273 - 7749520 on Build GRCh38
UniGene ID
Hs.75285
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002216.2 NP_002207.2 21 3113
GenBank mRNA AK225277.1 - 23 2976
AK225329.1 - 21 3018
X07173.1 - 21 3014

View Full Product Details