Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000068.3 | NP_000059.3 | 11 | 1614 | |
NM_001127221.1 | NP_001120693.1 | 11 | 1614 | ||
NM_001127222.1 | NP_001120694.1 | 11 | 1611 | ||
NM_001174080.1 | NP_001167551.1 | 11 | 1614 | ||
NM_023035.2 | NP_075461.2 | 11 | 1614 | ||
GenBank mRNA | AB035727.2 | - | 11 | 1375 | |
AF004883.1 | - | 11 | 1614 | ||
AF004884.1 | - | 11 | 1614 | ||
BQ340521.1 | - | 4 | 242 | ||
FJ040507.1 | - | 11 | 1378 | ||
U79666.1 | - | 11 | 1595 | ||
X99897.1 | - | 11 | 1653 |