Gene Transcripts

Gene Symbol
ACAD10
Entrez Gene ID
80724
Gene Name
acyl-CoA dehydrogenase family member 10
Gene Aliases
Chromosome Location
Chr. 12: 111686053 - 111757107 on Build GRCh38
UniGene ID
Hs.331141
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001136538.1 NP_001130010.1 7 1086
NM_025247.5 NP_079523.3 6 993
GenBank mRNA AK092356.1 - 6 978
AL832043.1 - 7 1064
AY323912.1 - 6 949
BC003698.1 - 2 150
BC015056.1 - 6 965
BC126358.1 - 6 907 8992074

View Full Product Details