Gene Transcripts

Gene Symbol
SAMD8
Entrez Gene ID
142891
Gene Name
sterile alpha motif domain containing 8
Gene Aliases
HEL-177, SMSr
Chromosome Location
Chr. 10: 75099592 - 75182123 on Build GRCh38
UniGene ID
Hs.744986
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144660.2 NP_653261.1 5 1105
GenBank mRNA AK057811.1 - 5 1033
EU668332.1 - 5 1033
GQ472208.1 - 5 1033

View Full Product Details