Gene Transcripts
- Gene Symbol
- SLC13A2
- Entrez Gene ID
- 9058
- Gene Name
- solute carrier family 13 member 2
- Gene Aliases
- NADC1, NaCT, NaDC-1, SDCT1
- Chromosome Location
- Chr. 17: 28473358 - 28497781 on Build GRCh38
- UniGene ID
- Hs.102307
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001145975.1 |
NP_001139447.1
|
4, 5 |
792 |
|
NM_003984.3 |
NP_003975.1
|
4, 5 |
645 |
|
NR_027384.1 |
|
5, 6 |
786 |
|
XM_006722165.2 |
XP_006722228.1
|
5, 6 |
694 |
|
XM_011525450.2 |
XP_011523752.1
|
4, 5 |
1058 |
|
XM_011525451.1 |
XP_011523753.1
|
5, 6 |
854 |
|
XM_011525452.1 |
XP_011523754.1
|
4, 5 |
645 |
|
XM_011525453.2 |
XP_011523755.1
|
4, 5 |
1066 |
|
XM_011525454.2 |
XP_011523756.1
|
4, 5 |
1080 |
|
GenBank mRNA |
AK091926.1 |
- |
6, 7 |
756 |
|
AK298388.1 |
- |
4, 5 |
700 |
|
AK303909.1 |
- |
5, 6 |
694 |
|
AK303936.1 |
- |
3, 4 |
424 |
|
AK314684.1 |
- |
4, 5 |
645 |
|
BC096276.1 |
- |
5, 6 |
740 |
40006564 |
BC096277.1 |
- |
4, 5 |
599 |
40006565 |
BC096278.1 |
- |
4, 5 |
599 |
40006566 |
BC096279.1 |
- |
4, 5 |
599 |
40006572 |
DB204108.1 |
- |
3, 4 |
424 |
|
U26209.1 |
- |
4, 5 |
646 |
|
View Full Product Details