Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_007284.3 | NP_009215.1 | 7 | 777 | |
GenBank mRNA | AF109365.1 | - | 7 | 678 | |
AL136773.1 | - | 7 | 744 | ||
BC000327.2 | - | 7 | 728 | 2822004 | |
BC003161.1 | - | 7 | 694 | 2957960 | |
BC016452.1 | - | 7 | 744 | 3452300 | |
CR456732.1 | - | 7 | 626 | ||
CR533520.1 | - | 7 | 626 | ||
Y17169.1 | - | 7 | 730 |