Gene Transcripts

Gene Symbol
ACOXL
Entrez Gene ID
55289
Gene Name
acyl-CoA oxidase-like
Gene Aliases
Chromosome Location
Chr. 2: 110732539 - 111118571 on Build GRCh38
UniGene ID
Hs.253320
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011511416.2 XP_011509718.1 19 2517
XM_011511419.2 XP_011509721.1 18 2427
GenBank mRNA AK001904.1 - 13 1776
BC131514.1 - 12 1683

View Full Product Details