Gene Transcripts
- Gene Symbol
- MORC1
- Entrez Gene ID
- 27136
- Gene Name
- MORC family CW-type zinc finger 1
- Gene Aliases
- CT33, MORC, ZCW6
- Chromosome Location
- Chr. 3: 108958239 - 109118146 on Build GRCh38
- UniGene ID
- Hs.591296
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_014429.3 |
NP_055244.3
|
10, 11 |
978 |
|
XM_005247362.2 |
XP_005247419.1
|
10, 11 |
935 |
|
XM_011512691.1 |
XP_011510993.1
|
10, 11 |
941 |
|
XM_011512692.1 |
XP_011510994.1
|
10, 11 |
941 |
|
XM_011512693.1 |
XP_011510995.1
|
10, 11 |
941 |
|
XM_011512694.1 |
XP_011510996.1
|
7, 8 |
692 |
|
XM_011512696.2 |
XP_011510998.1
|
4, 5 |
392 |
|
XM_011512697.2 |
XP_011510999.1
|
3, 4 |
560 |
|
XM_017006169.1 |
XP_016861658.1
|
10, 11 |
941 |
|
XM_017006170.1 |
XP_016861659.1
|
3, 4 |
360 |
|
GenBank mRNA |
AF084946.1 |
- |
10, 11 |
953 |
|
AK302642.1 |
- |
10, 11 |
935 |
|
AL353955.1 |
- |
4, 5 |
338 |
|
BC028680.1 |
- |
10, 11 |
979 |
|
BC050307.1 |
- |
10, 11 |
917 |
4838254 |
View Full Product Details