Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001320595.1 | NP_001307524.1 | 10 | 1484 | |
NM_001320596.1 | NP_001307525.1 | 10 | 1373 | ||
NM_001320597.1 | NP_001307526.1 | 9 | 1224 | ||
NM_004396.4 | NP_004387.1 | 9 | 1459 | ||
GenBank mRNA | AB209919.1 | - | 9 | 1211 | |
AB451257.1 | - | 9 | 1057 | ||
AB451382.1 | - | 9 | 1057 | ||
AK297192.1 | - | 8 | 989 | ||
AK297753.1 | - | 10 | 1194 | ||
AK297905.1 | - | 8 | 1017 | ||
AK310318.1 | - | 9 | 1103 | ||
BC016027.1 | - | 9 | 1198 | 3528578 | |
BT006943.1 | - | 9 | 1057 | ||
BX571764.1 | - | 9 | 1228 | ||
X15729.1 | - | 9 | 1224 | ||
X52104.1 | - | 9 | 1232 |