Gene Transcripts
- Gene Symbol
- DDX21
- Entrez Gene ID
- 9188
- Gene Name
- DEAD-box helicase 21
- Gene Aliases
- GUA, GURDB, RH-II/GU, RH-II/GuA
- Chromosome Location
- Chr. 10: 68956123 - 68985069 on Build GRCh38
- UniGene ID
- Hs.223141
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001256910.1 |
NP_001243839.1
|
7, 8 |
1466 |
|
NM_004728.3 |
NP_004719.2
|
7, 8 |
1347 |
|
XM_011540336.1 |
XP_011538638.1
|
7, 8 |
1339 |
|
XM_017016910.1 |
XP_016872399.1
|
7, 8 |
1257 |
|
GenBank mRNA |
AK025629.1 |
- |
4, 5 |
649 |
|
AK315585.1 |
- |
7, 8 |
1296 |
|
BC008071.2 |
- |
7, 8 |
1285 |
2987980 |
BX648405.1 |
- |
7, 8 |
1233 |
|
CR749598.1 |
- |
7, 8 |
1467 |
|
U41387.1 |
- |
7, 8 |
1298 |
|
View Full Product Details